missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Safe-Lock Tubes
GREENER_CHOICE

The hinged lid of Eppendorf Safe-Lock Tubes offers outstanding protection against unintentional opening, and completely eliminates any danger of leaks during incubation and storage.

Brand:  Eppendorf™ 0030121120

56.57 GBP valid until 2025-07-11
Use promo code "25721" to get your promotional price.


Product Code. 10394591

  • £61.90 / Pack of 500
Estimated Shipment: 30-05-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Hinged Eppendorf Safe-Lock lid prevents unintentional lid opening during incubation and storage
  • Polypropylene provides high resistance to chemicals, mechanical stress and temperature extremes
  • Manufactured without the use of slip agents, plasticizers, and biocides – substances that have been shown to leach from plastic consumables into the sample and negatively affect bio-assay results
  • g-Safe™: Depending on Safe-Lock Tube variant (see manual) exceptional centrifugation stability up to 30.000 × g prevents sample loss due to tube breakage and provides extra safety when working with hazardous samples
    - Precise lid sealing for minimal evaporation rates during long-term storage
  • Large frosted lid and surface on the side for easy labeling
  • Ensured functionality from -86 °C to 100 °C
  • Autoclavable when open (121 °C, 20 min)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

0.5 mL
500
-86 – 100°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Eppendorf™ Safe-Lock Tubes > Capacity: 0.5 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Eppendorf Safe-Lock Tubes are single use sample tubes intended as primary vessel in accordance with EN 14254:2004 and for storing human test specimen, with the exception of blood samples for in vitro diagnostics. Eppendorf Safe-Lock Tubes are intended for use by trained and skilled personnel only.