Learn More
Eppendorf™ Safe Lock Micro Test Tube

Use these polypropylene micro tubes for a variety of applications. Eppendorf™ Safe Lock Micro Test Tubes feature hinged Eppendorf Safe-Lock lids for outstanding protection against unintentional opening during incubation and storage.
Brand: Eppendorf™ 0030121139
58.69 GBP valid until 2025-05-31
Use promo code "25977" to get your promotional price.
Description
- Hinged Eppendorf Safe-Lock lid prevents unintentional lid opening during incubation and storage
- Polypropylene provides high resistance to chemicals, mechanical stress and temperature extremes
- Manufactured without mold release agents, softeners or biocides that could have a negative effect on bioassays
- g-Safe™: Exceptional centrifugation stability up to 30,000 × g prevents sample loss due to tube breakage and provides extra safety when working with hazardous samples
- Precise lid sealing for minimal evaporation rates during long-term storage
- Large frosted lid and surface on the side for easy labeling
- Guaranteed functionality from -86 to +100°C
- Autoclavable when open (121°C, 20 min.)
- Available in lot-certified PCR clean, Eppendorf Forensic DNA Grade, and Biopur quality for guaranteed purity
- Available in Eppendorf Protein and DNA LoBind material for maximum recovery of valuable samples

Specifications
0.5 mL | |
Polypropylene | |
–86 to 100°C |
Blue | |
500/Pk. |
Product Suggestions
Customers who viewed this item also viewed.
Special Offers
-
Multiple Manufacturers
Back to the Lab Summer Semester
Save up to 94% on Teaching Lab EssentialsPromo Code: 25721Expires: 2025-05-01
New Offer Save Now
Your input is important to us. Please complete this form to provide feedback related to the content on this product.