missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Screw Cap Septum Vials

For a convenient, versatile, and cost-effective vial and closure system, choose Thermo Scientific™ Screw Cap Septum Vials.

Brand:  Thermo Scientific™ TS-13019

109.62 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 11547340

  • £119.00 / Pack of 72

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

For a convenient, versatile, and cost-effective vial and closure system, choose Thermo Scientific™ Screw Cap Septum Vials. Made of autoclavable borosilicate glass and available with a wide assortment of special closures and accessories, these vials are ideal for sample collection and small derivatization reactions for use with automated gas (GC) and liquid chromatography (LC), and for storage of reagents and standards under complete seal with instant syringe access. The heavy-duty, flip-top divider box provides easy access to vials, caps, and septa and offers a convenient sample storage center. Clear and amber vials are available.

Applications

The 40mL clear or amber vial with a Thermo Scientific Disc is suitable for discrete water sampling under U.S. EPA 40 CFR Parts 136 and 141.

The 1.5mL clear and amber vials fit GC and LC autosamplers that use standard 12 ´ 32 mm vials.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

3.5mL; Clear
15 mm
3.5 mL
Autoclavable
45 mm
Septum Vial
Clear
Borosilicate Glass
3.5 mL
Screw Cap
Certificate of Analysis (COA), Certificate of Conformance (COC)
13-425
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Screw Cap Septum Vials > 3.5mL; Clear

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.