Learn More
Applied Biosystems™ SeqStudio™ Cartridge v2

SeqStudio™ Cartridge v2 is an all-in-one, easy-to-use reagent cartridge that includes capillaries, universal polymer, buffer, and pump required to run on the SeqStudio Genetic Analyzer System. The universal polymer allows for flexibility to perform Sanger sequencing and fragment analysis on the same instrument, with the same cartridge.
Brand: Applied Biosystems™ A41331
1884.65 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Key features:
- Minimize hands-on setup time with all-in-one cartridge
- Track consumables usage through radio frequency identification (RFID)
- Extended on-instrument 6-month shelf life for cartridge after opening
- Allows for 250 injections or 1000 reactions per cartridge*
- Responsible, simple, no-charge recycling program
* SeqStudio Cartridge v2 is optimized to work with the latest software version for the Seqstudio Genetic Analyzer. Please ensure that Seqstudio Genetic Analyzer is running the latest software before using this cartridge.

Specifications
Comparative Sequencing (Somatic Mutations - 10:50), De Novo Sequencing - Low to Mid Throughput, Fragment Analysis (Denaturing), Rare Variant Sequencing, Resequencing, Short PCR Product Sequencing, Short Read Sequencing | |
Capillaries, Universal polymer, Buffer, Pump | |
250 injections/1000 reactions | |
Capillary length: 28 cm |
SeqStudio Genetic Analyzer System | |
1000 | |
Store in refrigerator (2 to 8°C) |
Product Suggestions
Customers who viewed this item also viewed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.