missing translation for 'onlineSavingsMsg'
Learn More
Silica gel 60, 0.036-0.071mm (215-400 mesh), Thermo Scientific Chemicals Product Code.: 11416810

Silica gel 60, 0.036-0.071mm (215-400 mesh), Thermo Scientific Chemicals

Product Code. 11416810
2500 g
Click to view available options
Quantity:
500 g
1000 g
2500 g
This item is not returnable. View return policy

Product Code. 11416810

Brand: Thermo Scientific Alfa Aesar L14004.0E

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Silica gel is used in column chromatography without applied pressure (gravity pulled). It is used as an adsorbent in various applications and in removing impurities. It is used as stationary base in column chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Silica gel is used in column chromatography without applied pressure (gravity pulled). It is used as an adsorbent in various applications and in removing impurities. It is used as stationary base in column chromatography.

Solubility
Soluble in hydrofluoric acid. Insoluble in water.

Notes
Keep the container tightly sealed. Store in cool, dry conditions in a well sealed container. Incompatible with hydrogen fluoride.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7631-86-9
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
PubChem CID 24261
ChEBI CHEBI:30563
IUPAC Name dioxosilane
SMILES O=[Si]=O

Specifications

Material Silica Gel
Quantity 2500 g
Mesh Size 215 to 400 mesh
Merck Index 14,8493
Odor Odorless
Solubility Information Soluble in hydrofluoric acid. Insoluble in water.
Chemical Name or Material Silica gel 60, 0.036 to 0.071mm

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10249470 Certificate TypeCertificate of Analysis Date25/01/2024 Catalog NumberL14004.0B,L14004.0E,L14004.36
Lot Number10239372 Certificate TypeCertificate of Analysis Date24/02/2023 Catalog NumberL14004.0B,L14004.0E,L14004.36
Lot Number10215449 Certificate TypeCertificate of Analysis Date30/01/2023 Catalog NumberL14004.0B,L14004.0E,L14004.36
Lot NumberA10Z006 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL14004.0B,L14004.0E,L14004.36
Lot Number10193165 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL14004.0B,L14004.0E,L14004.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Silica gel 60, 0.036-0.071mm (215-400 mesh), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.