missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SiteClick™ Antibody Labeling Kits

Generate brightly labeled antibodies for flow cytometry, fluorescence imaging, or western blot detection using SiteClick™ Qdot™ R-PE, and Biotin Antibody Labeling Kits.

Brand:  Invitrogen™ S10453

431.68 GBP valid until 2025-04-18
Use promo code "24107" to get your promotional price.


 View more versions of this product

Product Code. 15138735

  • £568.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Contains everything needed to azido modify and label 100 μg of antibody.
1 kit
405/655
100 μg
Qdot™, SiteClick™
Far-Red
Qdot 525-800
Labeling Kit
Store the entire kit at 2 to 6°C upon receipt
SiteClick Labeling System
Wet Ice
Qdot 655
IgG
UV, Violet, Blue, Green, Yellow
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2994105 Certificate TypeCertificate of Analysis Date25/10/2024 Catalog NumberS10453
Lot Number2920600 Certificate TypeCertificate of Analysis Date30/07/2024 Catalog NumberS10453
Lot Number2761158 Certificate TypeCertificate of Analysis Date16/03/2024 Catalog NumberS10453
Lot Number2610304 Certificate TypeCertificate of Analysis Date13/10/2023 Catalog NumberS10453
Lot Number2566190 Certificate TypeCertificate of Analysis Date14/05/2023 Catalog NumberS10453
Special Offers

Special Offers

  • Multiple Manufacturers
    Advance your proteomics research with essential tools
    Promo Code: 25977

    Expires2025-05-31

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ SiteClick™ Antibody Labeling Kits > 1 kit

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.