missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Dopamine D1-like receptor agonist. Centrally active following systemic administration in vivo.

Specifications
Specifications
Quantity | 10 mg |
Synonym | skf 81297 hydrobromide, +/--6-chloro-pb hydrobromide, 6-chloro-1-phenyl-2,3,4,5-tetrahydro-1h-3-benzazepine-7,8-diol hydrobromide, y-6-chloro-pb hydrobromide, ?-6-chloro-pb hydrobromide, inverted question mark-6-chloro-pb hydrobromide, inverted question mark-skf-81297 hydrobromide, inverted exclamation marka-skf-81297 hydrobromide, inverted exclamation marka-6-chloro-pb hydrobromide, inverted exclamation marka-skf 81297 hydrobromide |
Formula Weight | 370.67 |
Percent Purity | >98% |
Chemical Name or Material | SKF 81297 hydrobromide |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SKF 81297 hydrobromide, Tocris Bioscience™ >
Spot an opportunity for improvement?Share a Content Correction