missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Potent and selective α2 antagonist at pre- and post-junctional α2-adrenoceptors (Ki values are 13 and 17 nM respectively). Exhibits antihypertensive activity in a rat model.

Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 232.15 |
Percent Purity | >98% |
Chemical Name or Material | SKF 86466 hydrochloride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SKF 86466 hydrochloride, Tocris Bioscience™ >
Spot an opportunity for improvement?Share a Content Correction