Learn More
Scientific Glass Labs™ Soda Lime Glass Watch Glasses
Manufactured from Soda lime glass. Scientific Glass Labs™ Soda Lime Glass Watch Glasses with thin, clear glass and ground edge can be used to evaporate liquids, hold solids for weighing, or for heating small substances.
Brand: Scientific Glass Labs™ 170/1/150
Description
Watch glasses are useful for evaporating liquids, holding solids for weighing, or for heating small substances.- Manufactured from Soda lime glass
- Thin, clear glass
- Ground edge
Specifications
Flat | |
Round | |
Evaporating Liquids, Heating Small Substances, Holding Solids for Weighing | |
150 mm |
Soda Lime Glass | |
Watch Glass | |
Ground Edge |