Learn More
Sodium Bis(2-methoxyethoxy)aluminumhydride, 70 wt. % Sol. in Toluene (ca. 3.5M), Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | Amber to Colorless |
Boiling Point | 110.0°C |
Linear Formula | [(CH3OCH2CH2O)2AlH2]Na |
Fieser | 02,382; 03,260; 04,441; 05,596; 06,528; 07,327; 08,448; 12,440; 15,290 |
Density | 1.0360g/mL |
Solubility Information | Solubility in water: reacts. Other solubilities: soluble in aromatic hydrocarbons and ethers |
Formula Weight | 202.16 |
Percent Purity | 68.5 to 71.5% |
Packaging | Glass bottle |
Specific Gravity | 1.036 |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0465828 | Certificate of Analysis | 12/09/2024 | 187900010,187902500,187900500 |
A0460170 | Certificate of Analysis | 20/02/2024 | 187900010,187902500,187900500 |
A0448561 | Certificate of Analysis | 11/12/2023 | 187900010,187902500,187900500 |
A0393947 | Certificate of Analysis | 15/12/2022 | 187900010,187902500,187900500 |
A0436440 | Certificate of Analysis | 21/11/2022 | 187900010,187902500,187900500 |
Safety and Handling




- Sodium bis(2-methoxyethoxy)aluminiumhydride, 70 wt% sol. in toluene (ca. 3.5M)
- Danger
- Aspiration toxicity Category 1
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
- Substance which\, in contact with water\, emits flammable gases Category 1
- H304-May be fatal if swallowed and enters airways.
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H361-Suspected of damaging fertility or the unborn child.
- H373-May cause damage to organs.
- H336-May cause drowsiness or dizziness.
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- H225-Highly flammable liquid and vapour.
- EUH014-Reacts violently with water.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P402+P404-Store in a dry place. Store in a closed container.
Special Offers
-
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
Days
31Hours
17Minutes
14Seconds
17New Offer Save Up to 70%
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.