missing translation for 'onlineSavingsMsg'
Learn More
Sodium ethoxide, 96%, Thermo Scientific Chemicals Product Code.: 11352426

Sodium ethoxide, 96%, Thermo Scientific Chemicals

Product Code. 11352426
500 g
Click to view available options
Quantity:
100 g
500 g
2000 g
This item is not returnable. View return policy

Product Code. 11352426

Brand: Thermo Scientific Alfa Aesar L06230.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sodium ethoxide is used as a strong base in organic synthesis. It is actively involved in the Claisen condensation, Stobbe reaction and Wolf-kishner reduction. It is an important starting material for the synthesis of ethyl ester and diethyl ester of malonic acid. In Williamson ether synthesis, it reacts with ethyl bromide to form diethyl ether. It is widely used as a base for the generation of carbanions which are utilized for alkylation and condensation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium ethoxide is used as a strong base in organic synthesis. It is actively involved in the Claisen condensation, Stobbe reaction and Wolf-kishner reduction. It is an important starting material for the synthesis of ethyl ester and diethyl ester of malonic acid. In Williamson ether synthesis, it reacts with ethyl bromide to form diethyl ether. It is widely used as a base for the generation of carbanions which are utilized for alkylation and condensation reactions.

Solubility
Soluble in ethanol and methanol.

Notes
Moisture sensitive. It undergoes hydrolysis rapidly to form sodium hydroxide in the presence of moist air. Incompatible with water, strong bases, acids, oxidizing agents, alkali metals, strong oxidizing agents, ammonia, acid chlorides, acid anhydrides, reducing agents, peroxides, acids and chlorinated solvents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-52-6
Molecular Formula C2H5NaO
Molecular Weight (g/mol) 68.051
MDL Number MFCD00012417
InChI Key QDRKDTQENPPHOJ-UHFFFAOYSA-N
Synonym sodium ethoxide, sodium ethylate, sodium ethanolate, sodiumethoxide, ethoxysodium, ethanol, sodium salt, caustic alcohol, naoet, etona, ethanol, sodium salt 1:1Show MoreShow Less
PubChem CID 2723922
ChEBI CHEBI:52096
IUPAC Name sodium;ethanolate
SMILES CC[O-].[Na+]

Specifications

Melting Point >300°C
pH 13
Flash Point 30°C (86°F)
Odor Irritating
Quantity 500 g
UN Number UN3206
Beilstein 3593646
Sensitivity Air and moisture sensitive
Merck Index 14,8613
Solubility Information Soluble in ethanol and methanol.
Formula Weight 68.05
Percent Purity 96%
Chemical Name or Material Sodium ethoxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0461024 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberL06230.0P,L06230.22,L06230.36
Lot Number10241345 Certificate TypeCertificate of Analysis Date20/02/2024 Catalog NumberL06230.0P,L06230.22,L06230.36
Lot NumberA0456515 Certificate TypeCertificate of Analysis Date17/10/2023 Catalog NumberL06230.0P,L06230.22,L06230.36
Lot Number10227111 Certificate TypeCertificate of Analysis Date13/10/2022 Catalog NumberL06230.0P,L06230.22,L06230.36
Lot Number10223921 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL06230.0P,L06230.22,L06230.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium ethoxide, 96%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.