Learn More
Nickel Electro™ Stainless Steel Flask Clip
For personalised clip arrangement, clips are supplied with all necessary fixings to enable them to be attached to the Perforated Tray. Nickel Electro™ Stainless Steel Flask Clip is suitable for flasks and can be autoclaved.
Brand:  Nickel Electro™ FC2000
 View more versions of this product
Product Code. 11776577
  • £31.30 / Each
Estimated Shipment: 30-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Highly flexible elastic design allowing glassware to easily slide into the clip and yet remain secure during mixing
  • No sharp edges to cut or score glassware
  • Can be autoclaved
  • Plastic base
  • Easy to assemble and remove Flask Clips from Perforated tray
Specifications
Specifications
2 L Conical Flask
Flask Clip
Stainless Steel
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Nickel Electro™ Stainless Steel Flask Clip > For Use With: 2000ml Conical Flask