Learn More
asecos™ Stainless Steel Safety Canister
The Stainless Steel Safety Canister is ideal for the safe distribution and dosing of flammable liquids.
Brand: asecos™ 36484
Description
- Incl. multilingual safety sticker for content labelling, with filling tap made of stainless steel (1.4301)
- Incl. ventilation, NBR seals and aluminium screw cap integrated flame arrester and plastic protective ring
Specifications
2.6 gal. | |
Gray | |
6.69 in. | |
11.02 in. | |
6.61 lb. |
10 L | |
Stainless Steel | |
170.00 mm | |
280.00 mm | |
3 kg |