Learn More
R&D Systems™ StemXVivo Neural Progenitor Differentiation Kit
Directed differentiation of human pluripotent stem cells into multipotent neural progenitor cells.
Brand: R&D Systems™ SC035
Description
Pluripotent stem cells (PSCs) are functionally defined by their capacity for self-renewal and their ability to differentiate into cells from all three germ layers. Mouse PSCs are characterized by their marker expression, including positive markers Oct-3/4, SSEA-1, Alkaline Phosphatase, and negative marker, SSEA-4. Optimal expansion and maintenance of mouse PSCs in culture is facilitated by using specialized stem cell base media supplemented with mouse leukemia inhibitory factor (LIF).
Benefits:
- Yields highly homogenous NPC population
- Kit-derived NPCs can be differentiated into astrocytes, oligodendrocytes, and multiple neuron subtypes
Storage: Store the unopened product at -20° to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Alternate Names: Pluripotent Stem Cells
Specifications
NPC Differentiation Base Media Supplement (100X), NPC Differentiation Cocktail (1000X), Anti-Human SOX1 Antibody. Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. | |
Sterile | |
RUO |
Stem Cell Differentiation Media | |
1 Kit | |
Embryonic Stem cells |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only