Learn More
Thermo Scientific™ Sterilin™ Weighing Boats
Form a funnel for easy pouring and complete sample transfer using the new and improved Thermo Scientific™ Sterilin™ Weighing Boats, featuring radiused corners.
£28.78 - £310.00
Product Code | Brand | Color | Capacity (Metric) | Type | Shape | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Capacity (Metric) | Type | Shape | Price | Quantity & Availability | ||
11864722
|
Thermo Scientific™
WB30332 |
Black | 5 mL | Antistatic Weighing Dish | Diamond |
£52.10
Pack of 500 |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||
11864712
|
Thermo Scientific™
WB30331 |
White | 5 mL | Antistatic Weighing Dish | Diamond |
£45.50
Pack of 500 |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||
11894712
|
Thermo Scientific™
WB30321 |
White | 7 mL | Weighing Boat | Square |
£93.20
Pack of 500 |
Estimated Shipment: 16-08-2024
Log in to see stock. |
||
11844712
|
Thermo Scientific™
WB30322 |
Black | 7 mL | Antistatic Weighing Dish | Square |
£55.62
Pack of 500 |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||
11333814
|
Thermo Scientific™
WB30205 |
White | 7 mL | Standard | Square |
£53.85
Pack of 500 |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||
12407702
|
Thermo Scientific™
WB30206 |
Black | 7 mL | Standard | Square |
£49.95
Pack of 500 |
Estimated Shipment: 20-11-2024
Log in to see stock. |
||
11894702
|
Thermo Scientific™
WB30305 |
Black | 250 mL | Standard | Square |
£130.00
Pack of 500 |
Estimated Shipment: 23-10-2024
Log in to see stock. |
||
11804712
|
Thermo Scientific™
WB30314 |
White | 30 mL | Standard | Diamond |
£220.00
Pack of 500 |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||
11824722
|
Thermo Scientific™
WB30315 |
Black | 30 mL | Standard | Diamond |
£63.50
Pack of 500 |
Estimated Shipment: 22-08-2024
Log in to see stock. |
||
11874712
|
Thermo Scientific™
WB30334 |
White | 30 mL | Antistatic Weighing Dish | Diamond |
£216.00
Pack of 500 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
||
11844722
|
Thermo Scientific™
WB30335 |
Black | 30 mL | Antistatic Weighing Dish | Diamond |
£88.75
Pack of 500 |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||
12437702
|
Thermo Scientific™
WB30317 |
White | 100 mL | Standard | Diamond |
£151.00
Pack of 500 |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||
11834722
|
Thermo Scientific™
WB30318 |
Black | 100 mL | Standard | Diamond |
£129.00
Pack of 500 |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||
12447702
|
Thermo Scientific™
WB30325 |
Black | 100 mL | Antistatic Weighing Dish | Square |
£135.00
Pack of 500 |
Estimated Shipment: 11-09-2024
Log in to see stock. |
||
11884712
|
Thermo Scientific™
WB30337 |
White | 100 mL | Antistatic Weighing Dish | Diamond |
£119.00
Pack of 500 |
Estimated Shipment: 11-09-2024
Log in to see stock. |
||
11804722
|
Thermo Scientific™
WB30324 |
White | 100 mL | Antistatic Weighing Dish | Square |
£159.00
Pack of 500 |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||
12417702
|
Thermo Scientific™
WB30255 |
Black | 100 mL | Standard | Square |
£71.95
Pack of 500 |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||
11854722
|
Thermo Scientific™
WB30338 |
Black | 100 mL | Antistatic Weighing Dish | Diamond |
£191.95
Pack of 500 |
Estimated Shipment: 22-08-2024
Log in to see stock. |
||
11824712
|
Thermo Scientific™
WB30254 |
White | 100 mL | Standard | Square |
£84.95
Pack of 500 |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||
11854712
|
Thermo Scientific™
WB30328 |
Black | 250 mL | Antistatic Weighing Dish | Square |
£168.00
Pack of 500 |
Estimated Shipment: 16-09-2024
Log in to see stock. |
||
11814722
|
Thermo Scientific™
WB30327 |
White | 250 mL | Antistatic Weighing Dish | Square |
£265.00
Pack of 500 |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||
11834712
|
Thermo Scientific™
WB30304 |
White | 250 mL | Standard | Square |
£310.00
Pack of 500 |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||
12427702
|
Thermo Scientific™
WB30311 |
White | 5 mL | Standard | Diamond |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
Beschreibung
Form a funnel for easy pouring and complete sample transfer using the new and improved Thermo Scientific™ Sterilin™ Weighing Boats, featuring radiused corners. All sizes packed 250 per pack, 500 per case.
- Manufactured from high impact polystyrene
- Hydrophobic surface ensures no absorption of water from atmosphere or sample
- Available in black or white to ensure suitability for different colored sample materials
- Anti-static range alleviates issues with powder hang-up
- Flat base for a secure, firm rest on the balance pan
- Supplied nonsterile
Spezifikation
Flat | |
Polystyrene |