Learn More
Cole Parmer Stopper Hole Boring Set
Chrome-plated steel cork borer set stays sharp.
Brand:  Cole Parmer 06298-90
Product Code. 15380367
  • £306.80 / Each
Estimated Shipment: 20-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Chrome-plated steel cork borer set stays sharp.
  • Flat blades permit easy boring of lightweight plastic, cork stoppers, and spongy articles.
  • Set consists of twelve borers with outside diameters of 4, 6, 7.5, 8.5, 10, 12, 13, 15, 17, 18.5, 19, and 20.5 mm.
Specifications
Specifications
10 mm, 12 mm, 13 mm, 15 mm, 17 mm, 18.5 mm, 19 mm, 20.5 mm, 4 mm, 6 mm, 7.5 mm, 8.5 mm
Chrome
Stopper Hole Boring Set
12 Borers/Set
Chrome Plated Steel
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cole Parmer Stopper Hole Boring Set > boring set