Learn More
Thermo Scientific™ SuperSignal™ West Atto Ultimate Sensitivity Substrate
SuperSignal™ West Atto Ultimate Sensitivity Substrate is an enhanced chemiluminescent (ECL) substrate for high attogram protein detection and provides the high level sensitivity and better signal-to-noise ratios compared to other commercially available horseradish peroxidase (HRP) substrates.
Brand: Thermo Scientific™ A38554
119.70 GBP valid until 2025-05-31
Use promo code "25977" to get your promotional price.
Description
- Sensitivity: low-femtogram to high-attogram
- Stability: eight hours for working solution, one year for kit at 4°C
- Compatibility: nitrocellulose and PVDF membranes
- Signal duration: up to four hours
- Recommended primary antibody concentration: 1:1000 to 1:5000 dilution (0.2 to 1.0 μg/mL)
- Recommended secondary antibody concentration: 1:100000 to 1:250000 dilution (4 to 10 ng/mL)

Specifications
Substrate | |
Liquid | |
Nitrocellulose, PVDF | |
20 mL | |
Low-femtogram to high-attogram | |
SuperSignal West Atto Substrate / SuperSignal West Atto Stable Peroxide • Substrate Solution, 10 mL • Stable Peroxide Solution, 10 mL Sufficient for two mini-gel blots; 200 cm2 of membrane. Store reagents at 2–8°C. |
Chemiluminescence | |
Western Blot | |
SuperSignal™ | |
Horseradish Peroxidase | |
2 Mini-Gel Blots, 200 sq. cm of Membrane | |
Wet Ice |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
ZI397786A | Certificate of Analysis | 15/10/2024 | A38554 |
ZI397786 | Certificate of Analysis | 15/10/2024 | A38554 |
ZF392528 | Certificate of Analysis | 21/06/2024 | A38554 |
ZF392528A | Certificate of Analysis | 21/06/2024 | A38554 |
ZC388323 | Certificate of Analysis | 12/04/2024 | A38554 |
Special Offers
-
Multiple Manufacturers
Powering Proteomics
Advance your proteomics research with essential toolsPromo Code: 25977Expires: 2025-05-31
New Offer Save Up to 92%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.