Learn More
PURUS™ Synthetic Cleanroom Paper
Cleanroom paper that excels in Tensile Strength. For writing, printing and photocopying in cleanroom environments, PURUS™ Cleanroom Paper provides excellent printability and heat toner adhesion and is recyclable.
Brand:  PURUS™ LCIB 2044C
171.95 GBP valid until 2025-05-02
Use promo code "24151" to get your promotional price.
 View more versions of this product
Product Code. 16450041
  • £181.00 / Pack of 1250
Estimated Shipment: 24-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Features high tensile strength and excellent heat resistance for enhanced copier and printing efficiency
  • Propriatary binding technology results in very low levels of particles, nonvolatile residue (NVR) and ion extractables
  • Unique precision cutting technology guarantees consistent sizing and cleanliness
  • Produced in ISO 4 (Class 10) cleanroom
  • Recyclable
  • Suitable for use in ISO 4 to 5 (Class 10 to Class 1000) cleanrooms
Specifications
Specifications
Synthetic
Blue
Autoclavable
Yes
250 Sheets/Pk., 5 Packs/Cs.
100 gsm
No
420 x 297 mm
For Class 10 to Class 1000 cleanrooms, writing, printing and photocopying in clean environments
Cleanroom Paper
A3
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
PURUS™ Synthetic Cleanroom Paper > Blue,A3,100gsm