Learn More
Applied Biosystems™ TaqMan™ Advanced miRNA Human A 96-well Plates, standard
The Applied Biosystems TaqMan Advanced miRNA Human A 96-Well Plates, Standard, provide the convenience of pre-spotted TaqMan Advanced miRNA assays in a 96-well standard plate format.
Brand: Applied Biosystems™ A31811
Description
This product contains a set of 4 plates for profiling of up to 376 unique miRNAs, and includes endogenous and exogenous miRNA controls for normalization of data results.
Features of the TaqMan Advanced miRNA Human A 96-Well Plates include:
- Reproducibility—pre-spotted assays are designed to reduce pipetting error and minimize experimental variability
- Sample savings—requires only 2 μL of purified plasma/serum or 1–10 ng of total RNA per sample
- Updated content—assay content updated for mirBase v.21
- Easy to use—just add a mix of diluted miR-Amp and master mix and run the qPCR; no additional pipetting required
- Gold-standard TaqMan qPCR chemistry—TaqMan Advanced miRNA assays draw on Thermo Fisher Scientific's industry-leading bioinformatics assay design pipeline and verification system to help ensure high specificity and minimal cross-reactivity, even for closely-related miRNA families
TaqMan Advanced miRNA plates require the use of the TaqMan Advanced cDNA Synthesis Kit, which uses a universal reverse transcription (RT) chemistry to prepare the cDNA template for use with TaqMan Advanced miRNA assays for detection and quantification of mature human miRNAs in biological samples.
These plates are not compatible with the first-generation TaqMan MicroRNA cDNA Synthesis Kit, which uses an miRNA-specific reverse transcription chemistry.
Specifications
Store at room temperature. | |
96-well plate | |
2-step RT-qPCR | |
7500 System, 7900HT System, QuantStudio™ 12k Flex, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7, ViiA™ 7 System, StepOnePlus™ System | |
Advanced miRNA Human 96-Well Plate | |
Room Temperature | |
Liquid Samples (e.g. Serum), Tissue, Cells, FFPE & Fixed Samples, Blood |
Primer-probe | |
High | |
Standard | |
TaqMan™ | |
4 plates | |
Microarray Analysis | |
2 μL |
Product Suggestions
Customers who viewed this item also viewed