Learn More
Retsch™ Test Sieves with Micro Mesh Sizes
Retsch™ Test Sieves consist of a solid stainless steel sieve frame with high stability, resulting in reliable sieving results.
£109.00 - £828.00
Specifications
Certifications/Compliance DIN ISO 3310-1
Material Stainless Steel
Includes Calibration Certificate
Height (Metric) 50 mm
Diameter (Metric) Outer 200 mm
View More Specs
Products 39
Product Code Brand Pore Size Price Quantity & Availability  
Product Code Brand Pore Size Price Quantity & Availability  
15654168
View Documents
Retsch™
60.131.000020
20 μm
£828.00
Each
Estimated Shipment: 10-07-2025
to see stock.
Add to basket
15655637
View Documents
Retsch™
60.131.000025
25 μm
£408.00
Each
Estimated Shipment: 07-05-2025
to see stock.
Add to basket
15665637
View Documents
Retsch™
60.131.000032
32 μm
£262.00
Each
Estimated Shipment: 14-05-2025
to see stock.
Add to basket
15675637
View Documents
Retsch™
60.131.000036
36 μm
£237.00
Each
Estimated Shipment: 10-07-2025
to see stock.
Add to basket
15600379
View Documents
Retsch™
60.131.000038
38 μm
£237.00
Each
Estimated Shipment: 10-07-2025
to see stock.
Add to basket
15500171
View Documents
Retsch™
60.131.000040
40 μm
£165.00
Each
Estimated Shipment: 23-06-2025
to see stock.
Add to basket
15685637
View Documents
Retsch™
60.131.000045
45 μm
£212.00
Each
Estimated Shipment: 03-06-2025
to see stock.
Add to basket
15590161
View Documents
Retsch™
60.131.000050
50 μm
£150.80
Each
Estimated Shipment: 14-05-2025
to see stock.
Add to basket
15610379
View Documents
Retsch™
60.131.000053
53 μm
£192.00
Each
Estimated Shipment: 23-06-2025
to see stock.
Add to basket
15674188
View Documents
Retsch™
60.131.000056
56 μm
£124.00
Each
In Stock
to see stock.
Edge
Add to basket
15665515
View Documents
Retsch™
60.131.000063
63 μm
£150.00
Each
Estimated Shipment: 14-05-2025
to see stock.
Add to basket
15510171
View Documents
Retsch™
60.131.000071
71 μm
£109.00
Each
Estimated Shipment: 10-07-2025
to see stock.
Add to basket
15613077
View Documents
Retsch™
60.131.000075
75 μm
£133.00
Each
Estimated Shipment: 14-05-2025
to see stock.
Add to basket
15640864
View Documents
Retsch™
60.131.000080
80 μm
£109.00
Each
Estimated Shipment: 10-07-2025
to see stock.
Add to basket
15695637
View Documents
Retsch™
60.131.000090
90 μm
£150.00
Each
Estimated Shipment: 14-05-2025
to see stock.
Add to basket
15580161
View Documents
Retsch™
60.131.000100
100 μm
£109.00
Each
Estimated Shipment: 14-05-2025
to see stock.
Add to basket
15575860
View Documents
Retsch™
60.131.000106
106 μm
£133.00
Each
Estimated Shipment: 22-05-2025
to see stock.
Add to basket
15620379
View Documents
Retsch™
60.131.000112
112 μm
£109.00
Each
Estimated Shipment: 10-07-2025
to see stock.
Add to basket
15630379
View Documents
Retsch™
60.131.000125
125 μm
£151.00
Each
Estimated Shipment: 14-05-2025
to see stock.
Add to basket
15605647
View Documents
Retsch™
60.131.000150
150 μm
£138.00
Each
Estimated Shipment: 14-05-2025
to see stock.
Add to basket
15590171
View Documents
Retsch™
60.131.000160
160 μm
£109.00
Each
Estimated Shipment: 10-07-2025
to see stock.
Add to basket
15640379
View Documents
Retsch™
60.131.000180
180 μm
£114.00
Each
Estimated Shipment: 29-05-2025
to see stock.
Add to basket
15530171
View Documents
Retsch™
60.131.000200
200 μm
£132.00
Each
Estimated Shipment: 14-05-2025
to see stock.
Add to basket
15650379
View Documents
Retsch™
60.131.000212
212 μm
£112.00
Each
Estimated Shipment: 14-05-2025
to see stock.
Add to basket
15660379
View Documents
Retsch™
60.131.000224
224 μm
£109.00
Each
Estimated Shipment: 10-07-2025
to see stock.
Add to basket
15550171
View Documents
Retsch™
60.131.000250
250 μm
£173.00
Each
Estimated Shipment: 03-06-2025
to see stock.
Add to basket
15670379
View Documents
Retsch™
60.131.000280
280 μm
£109.00
Each
Estimated Shipment: 10-07-2025
to see stock.
Add to basket
15680379
View Documents
Retsch™
60.131.000300
300 μm
£115.00
Each
Estimated Shipment: 07-05-2025
to see stock.
Add to basket
15560171
View Documents
Retsch™
60.131.000315
315 μm
£117.52
Each
Estimated Shipment: 05-06-2025
to see stock.
Add to basket
15690379
View Documents
Retsch™
60.131.000355
355 μm
£114.00
Each
Estimated Shipment: 10-07-2025
to see stock.
Add to basket
15600389
View Documents
Retsch™
60.131.000425
425 μm
£112.00
Each
Estimated Shipment: 10-07-2025
to see stock.
Add to basket
15610389
View Documents
Retsch™
60.131.000450
450 μm
£109.00
Each
Estimated Shipment: 10-07-2025
to see stock.
Add to basket
15580171
View Documents
Retsch™
60.131.000500
500 μm
£163.00
Each
Estimated Shipment: 14-05-2025
to see stock.
Add to basket
15620389
View Documents
Retsch™
60.131.000560
560 μm
£111.00
Each
Estimated Shipment: 10-07-2025
to see stock.
Add to basket
15630389
View Documents
Retsch™
60.131.000600
600 μm
£111.00
Each
Estimated Shipment: 10-07-2025
to see stock.
Add to basket
15578841
View Documents
Retsch™
60.131.000630
630 μm
£111.00
Each
Estimated Shipment: 10-07-2025
to see stock.
Add to basket
15588841
View Documents
Retsch™
60.131.000800
800 μm
£111.00
Each
Estimated Shipment: 10-07-2025
to see stock.
Add to basket
15640389
View Documents
Retsch™
60.131.000850
850 μm
£111.00
Each
Estimated Shipment: 23-06-2025
to see stock.
Add to basket
15650389
View Documents
Retsch™
60.131.000900
900 μm
£111.00
Each
Estimated Shipment: 10-07-2025
to see stock.
Add to basket
Description
Description
  • High quality joining technology guarantees permanently taut wire mesh
  • One-piece cast sieve frame and seamless mesh joint prevent cross contamination
  • Stainless steel sieve frame with high form stability
  • High degree of corrosion resistance and easy cleaning thanks to high-alloy stainless steel
  • Excellent product quality due to extensive optical inspection
  • Maximum stability and optimum sealing when used in sieve stacks thanks to the o-ring which is placed in the recess designed for this purpose
  • Clear and precise labeling of the sieves with full traceability based on individualized laser engraving
Specifications
Specifications
DIN ISO 3310-1
Calibration Certificate
200 mm
Stainless Steel
50 mm
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Retsch™ Test Sieves with Micro Mesh Sizes