Learn More
Thermo Scientific™ Three60™ Sampling System
Allows you to take multiple representative samples of your process liquids conveniently and in a closed system.
Brand: Thermo Scientific™ 4MP0037
Description
The Thermo Scientific™ Three60™ Sampling System allows you to take a representative sample of your product with minimal effort. The fluid paths of the four pre-irradiated BioProcess Containers (BPCs) and assemblies feature quick-turn valves and pinch-and-cut disconnections for fluid transfer and removal. The Three60 Sampling System is composed of Thermo Scientific™ ASI™ 77 film, a 5-layer, 7mL, polyethylene film and includes 2 ports (1 luer lock insert + cap, 1 injection site).
- Compatible with a 3.8 cm (1.5-inch) sanitary connector
- Requires no expensive hardware
- Each Three60 package contains a valve and four assemblies with pinch-and-cut disconnections
- Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
Applications
- Removes the need for tank cleaning and helps to reduce cycle times
- Facilitates hydration of powdered media and buffers, and pooling of non-sterile solutions and fluids

Specifications
500 mL | |
22.0 x 13.5 cm (8.66 x 5.32 in.) | |
Placed into a thermoform tray and sealed | |
Ethyl Vinyl Acetate | |
Room Temperature | |
Three60™ | |
Non-sterile |
Store at room temperature. | |
ASI™ 77 | |
2 Ports (1 Luer Lock Insert + Cap, 1 Injection Site) | |
8/Cs. | |
Irradiated | |
Sampling System |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.