Learn More
Cytiva Tricorn Storage/Shipping Device
Designed for high-resolution liquid chromatography of biomolecules performed at medium to high pressures.
Brand: Cytiva 18-1176-43
88.35 GBP valid until 2025-05-31
Use promo code "24127" to get your promotional price.
Description
- Suitable for use with high performance media
- Glass tubes coated with plastic film
Specifications
Glass tubes coated with plastic film | |
Sepharose High Performance, Superdex Prep Grade, and SOURCE | |
Tricorn Storage/Shipping Device |