missing translation for 'onlineSavingsMsg'
Learn More
Honeywell™ TRICOTON Light Gloves Product Code.: 12693343
In Stock
to see stock.
Add to basket

Honeywell™ TRICOTON Light Gloves

Product Code. 12693343
8
Click to view available options
Size:
7
8

Product Code. 12693343

Brand: Honeywell™ RGT550T8

In Stock
to see stock.
Add to basket
Add to basket

Honeywell™ TRICOTON Light Gloves are a 100% cotton lightweight knitted glove constructed of light yarn.

  • 100% cotton lightweight knitted glove
  • The knitting technique helps to obtain a glove that conforms snugly to the hand and good dexterity
  • The glove offers maximum comfort and added sturdiness
  • Of a neutral pH level and without any treatment, this cotton is tolerated perfectly well by the skin and limits the absorption of perspiration
  • The tight knit gives the glove perfect support, limits the penetration of dirt, offers a good touch while offering protection against small cuts
  • Light color enhances the glove's hygiene and cleanliness properties
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EN 420-2003 EEC Basic standards
Construction Seamless Knit
Description 13 gauge
Material Cotton
Size 8
Glove Cuff Style Elasticated
Length (Metric) 23 cm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Honeywell™ TRICOTON Light Gloves >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.