Learn More
Trimethylsulfoxonium iodide, 98+%, Thermo Scientific Chemicals
Description
Trimethylsulfoxonium iodide reacts with sodium hydride to prepare dimethyloxosulfonium methylide, which is used as a methylene-transfer reagent in synthetic chemistry. It is used to prepare ylide, which reacts with carbonyl compounds to get epoxides. Further, it reacts with alfa,beta-unsaturated esters to get cyclopropyl esters.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Trimethylsulfoxonium iodide reacts with sodium hydride to prepare dimethyloxosulfonium methylide, which is used as a methylene-transfer reagent in synthetic chemistry. It is used to prepare ylide, which reacts with carbonyl compounds to get epoxides. Further, it reacts with alfa,beta-unsaturated esters to get cyclopropyl esters.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents and strong bases. Light sensitive.

Specifications
Specifications
Melting Point | ∼170°C (decomposition) |
Quantity | 100 g |
Beilstein | 3595854 |
Sensitivity | Light sensitive |
Solubility Information | Soluble in water. |
Formula Weight | 220.07 |
Percent Purity | ≥98% |
Chemical Name or Material | Trimethylsulfoxonium iodide |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10251945 | Certificate of Analysis | 09/08/2024 | A14589.14,A14589.36,A14589.22 |
10209178 | Certificate of Analysis | 22/11/2021 | A14589.14,A14589.36,A14589.22 |
10097577 | Certificate of Analysis | 22/11/2021 | A14589.14,A14589.36,A14589.22 |
10167485 | Certificate of Analysis | 22/11/2021 | A14589.14,A14589.36,A14589.22 |
10161447 | Certificate of Analysis | 22/11/2021 | A14589.14,A14589.36,A14589.22 |
Safety and Handling

- Warning
- LONG-TERM AQUATIC HAZARD Chronic 2
- ACUTE AQUATIC Acute 1
- H411-Toxic to aquatic life with long lasting effects.
- H400-Very toxic to aquatic life.
- P273-Avoid release to the environment.
- P391-Collect spillage.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.