missing translation for 'onlineSavingsMsg'
Learn More
Trimethylsulfoxonium iodide, 98+%, Thermo Scientific Chemicals Product Code.: 11412008

Trimethylsulfoxonium iodide, 98+%, Thermo Scientific Chemicals

Product Code. 11412008
500 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11412008

Brand: Thermo Scientific Alfa Aesar A14589.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Trimethylsulfoxonium iodide reacts with sodium hydride to prepare dimethyloxosulfonium methylide, which is used as a methylene-transfer reagent in synthetic chemistry. It is used to prepare ylide, which reacts with carbonyl compounds to get epoxides. Further, it reacts with alfa,beta-unsaturated esters to get cyclopropyl esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trimethylsulfoxonium iodide reacts with sodium hydride to prepare dimethyloxosulfonium methylide, which is used as a methylene-transfer reagent in synthetic chemistry. It is used to prepare ylide, which reacts with carbonyl compounds to get epoxides. Further, it reacts with alfa,beta-unsaturated esters to get cyclopropyl esters.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents and strong bases. Light sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1774-47-6
Molecular Formula C3H9IOS
Molecular Weight (g/mol) 220.07
MDL Number MFCD00011899
InChI Key BPLKQGGAXWRFOE-UHFFFAOYSA-M
Synonym trimethylsulfoxonium iodide, trimethylsulphoxonium iodide, trimethyloxosulfonium iodide, sulfoxonium, trimethyl-, iodide, trimethyloxosulphonium iodide, trimethyl sulphoxonium ioidie, s,s,s-trimethylsulfoxonium iodide, trimethylsulfoxoniumiodide, sulfonium, trimethyl-, iodide, oxide, sulfoxonium, trimethyl-, iodide 1:1Show MoreShow Less
PubChem CID 74498
IUPAC Name trimethyl(oxo)-λ⁶-sulfanylium iodide
SMILES [I-].C[S+](C)(C)=O

Specifications

Melting Point ∼170°C (decomposition)
Quantity 500 g
Beilstein 3595854
Sensitivity Light sensitive
Solubility Information Soluble in water.
Formula Weight 220.07
Percent Purity ≥98%
Chemical Name or Material Trimethylsulfoxonium iodide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10251945 Certificate TypeCertificate of Analysis Date09/08/2024 Catalog NumberA14589.14,A14589.36,A14589.22
Lot Number10209178 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14589.14,A14589.36,A14589.22
Lot Number10097577 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14589.14,A14589.36,A14589.22
Lot Number10167485 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14589.14,A14589.36,A14589.22
Lot Number10161447 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14589.14,A14589.36,A14589.22
compliance-icons
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H411-Toxic to aquatic life with long lasting effects.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P391-Collect spillage.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Trimethylsulfoxonium iodide, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.