missing translation for 'onlineSavingsMsg'
Learn More
Trioctyl phosphate, Thermo Scientific Chemicals Product Code.: 11397138

Trioctyl phosphate, Thermo Scientific Chemicals

Product Code. 11397138
1 kg
Click to view available options
Quantity:
50 g
250 g
1 kg
This item is not returnable. View return policy

Product Code. 11397138

Brand: Thermo Scientific Alfa Aesar 015808.A1

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Trioctyl phosphate finds application as a fire-retardant, plasticizer for polyvinyl chloride and cellulose nitrate. It is used as a stimulant for the chemical warfare nerve agent VX. Further, it used as an alkylation agent for nitrogen heterocyclic compounds and as a catalyst to produce phenolic and urea resins. In addition to this, it is used as an antifoaming agent and a co-solvent in hydrogen peroxide production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-42-2
Molecular Formula C24H51O4P
Molecular Weight (g/mol) 434.64
MDL Number MFCD00009491
InChI Key GTVWRXDRKAHEAD-UHFFFAOYNA-N
Synonym tris 2-ethylhexyl phosphate, disflamoll tof, flexol tof, phosphoric acid, tris 2-ethylhexyl ester, kronitex tof, flexol plasticizer tof, 2-ethyl-1-hexanol phosphate, tris ethylhexyl phosphate, tri 2-ethylhexyl phosphateShow MoreShow Less
PubChem CID 6537
IUPAC Name tris(2-ethylhexyl) phosphate
SMILES CCCCC(CC)COP(=O)(OCC(CC)CCCC)OCC(CC)CCCC

Specifications

Density 0.93 g/mL
Boiling Point 216°C (4 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.441
Quantity 1 kg
Solubility Information Miscible with mineral oil and gasoline. Immiscible with water.
Formula Weight 434.64
Physical Form Liquid
Chemical Name or Material Trioctyl phosphate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberS14I023 Certificate TypeCertificate of Analysis Date27/06/2022 Catalog Number015808.A1,015808.30,015808.18
Lot NumberZ06F051 Certificate TypeCertificate of Analysis Date03/06/2022 Catalog Number015808.A1,015808.30,015808.18
Lot NumberE29U025 Certificate TypeCertificate of Analysis Date28/12/2021 Catalog Number015808.A1,015808.30,015808.18
Lot NumberL30W024 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number015808.A1,015808.30,015808.18
Lot NumberF01S024 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number015808.A1,015808.30,015808.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Trioctyl phosphate, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.