Learn More
Thermo Scientific™ Tryptone Soya Broth (Bottled)
Grow a wide variety of microprganisms using this medium in convenient ready prepared bottles.
Brand: Thermo Scientific™ BO0369E
63.03 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Grow a wide variety of microorganisms with Thermo Scientific™ Oxoid™ Tryptone Soya Broth ready-prepared bottled medium. The general purpose medium, containing two peptones, is suitable for the cultivation of both aerobes and anaerobes. It is specified in International Pharmacopoeia1,2,3,4 and food testing methods for the growth of a wide variety of organisms.
Remove the need for autoclaving and dispensing with convenient, pre-filled and ready-to-use bottles of Tryptone Soya Broth.
- Conforms to formulations detailed in International Pharmacopoeia.
- Convenient: pre-filled bottles are ready for immediate use.
- Simple: easy to use with minimum training.
- Long shelf life: up to 12 months.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

Specifications
Straw, Clear | |
Tryptone Soya Broth (EP/USP) (Bottle) | |
United Kingdom | |
10.0g + 0.3g | |
24/Pk. |
2 to 25°C | |
Vial Bottles | |
Microbiology Media | |
Twenty four vial bottles with narrow septum, injectable closures in a box |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
4496844 | Certificate of Analysis | 20/03/2025 | BO0369E |
4494484 | Certificate of Analysis | 11/03/2025 | BO0369E |
4494485 | Certificate of Analysis | 11/03/2025 | BO0369E |
4489673 | Certificate of Analysis | 25/02/2025 | BO0369E |
4488300 | Certificate of Analysis | 20/02/2025 | BO0369E |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.