Learn More
Cytiva UV Flow Cell
Flow cell design with fiber optics ensures high signal-to-noise ratio with minimal drift and refractive index effects.
Brand:  Cytiva 18-1111-10
 View more versions of this product
Product Code. 11340232
  • £1832.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Monitor used as standalone detector or as part of AKTAdesign system
  • UNF 10-32 fingertight connectors for capillary tubing with 0.062 in. outer dia.
  • Made from PEEK, PTFE, Ti alloy and glass
TRUSTED_SUSTAINABILITY
Specifications
Specifications
UV Flow Cell
Flow cell design with fiber optics ensures high signal-to-noise ratio with minimal drift and refractive index effects
PEEK, PTFE, Titanium Alloy, Glass
2 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cytiva UV Flow Cell >