Learn More
uvex i-3 Safety Goggles Product Code.: 15366062
In Stock
to see stock.
Add to basket
uvex i-3 Safety Goggles
Product Code. 15366062 Shop All Uvex Products
Black, Light Gray, Clear
Click to view available options
Frame Color:
Anthracite Blue
Black, Light Gray
Lens Tint:
Clear
Gray
Product Code. 15366062
Brand: uvex 9190280
In Stock
to see stock.
Edge
Add to basket
Add to basket
Innovative metal-free protective spectacles, designed to perfectly fit the face
  • A new dimension in protective eyewear - these innovative metal-free protective spectacles are designed to perfectly fit the face
  • Ergonomically designed Softflex bridge piece, soft and variable nose loops do not slip and sit comfortably on the nose, Softflex ear pieces, variable side-arm
  • Angle, adjustable side-arm flex zone
  • Coating uvex supravision excellence
  • TRUSTED_SUSTAINABILITY
    Specifications
    Certifications/Compliance EN 170
    Lens Type Uvex™ supravision excellence
    Lens Coating Anti-fog
    Description Shield-lens spectacle
    Lens Tint Clear
    Product Type Ergonomic Safety Glasses
    Frame Color Black, Light Gray
    Material Polycarbonate
    Disposable Reusable
    Lens Material Polycarbonate
    Frame Material Plastic
    UV Protection UV400
    Frame Type Standard
    Show More Show Less
    • Multiple Manufacturers
      Save up to 60% on gloves, apparel, and more
      Promo Code: 24131
      Expires2025-06-08
      New Offer
      Shop Now
    Product Content Correction
    Product Title
    uvex i-3 Safety Goggles >