missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Verti-Gel Mini Tetrad PAGE System

Durable and leakproof system to run up to four gels.

Brand:  Fisherbrand™ FCVS10TETRAD1

 View more versions of this product

Product Code. 15116624

  • ÂŁ539.00 / Each
Estimated Shipment: 08-07-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Injection-moulded construction
  • Compatible with all 80 x 100mm and 100 x 100mm precast gels
  • Low buffer volumes
Specifications

Specifications

Verti-Gel Mini Tetrad PAGE System
100 x 100mm sliding clamps, caster and external stand, four notched/plain plates with 1mm bonded spacers, four notched plates, four 12-well 1mm combs
4
250 to 1200 mL
75 x 80 mm
Injection-molded
0.5 mm, 0.75 mm, 1 mm, 1.5 mm, 2 mm combs
190 x 130 x 150 mm
Product Suggestions

Product Suggestions

Videos

Videos

  • Fisherbrand and Fisher Bioreagents - Focus on electrophoresis
    Fisherbrand provides a comprehensive range of horizontal gel systems for all your nucleic acid gel electrophoresis needs.
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
     
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected

      Fisherbrand and Fisher BioReagents Focus on electrophoresis

    • Fisherbrand - Meet the Family
      Fisher Scientific's private label product line, Fisherbrand™, offers customers a broad selection of laboratory supplies & consumables, covering a range of applications such as pH & electrochemistry, stirring & mixing, centrifugation, liquid handling,
      Video Player is loading.
      Current Time 0:00
      Duration 0:00
      Loaded: 0%
      Stream Type LIVE
      Remaining Time 0:00
       
      1x
      • Chapters
      • descriptions off, selected
      • captions off, selected

        Fisherbrand Meet the Family

      SDS
      Documents

      Documents

      Certificates
      Special Offers

      Special Offers

      Product Content Correction

      Your input is important to us. Please complete this form to provide feedback related to the content on this product.

      Product Title
      Fisherbrand™ Verti-Gel Mini Tetrad PAGE System >

      By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

      Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.