missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Vivaspin™ 20 Centrifugal Concentrator
BIOPHARMA

Offers increased volume flexibility and performance. VivaspinTM 20 Centrifugal Concentrator handles up to 20mL in swing bucket rotors and 14mL in fixed angle rotors accepting 50mL centrifuge tubes. Features twin vertical membranes for unparalleled filtration speed and >100-fold concentration.

Brand:  Sartorius VS2091

142.99 GBP valid until 2025-05-31
Use promo code "25977" to get your promotional price.


 View more versions of this product

Product Code. 10012561

  • £351.00 / Pack of 12
Estimated Shipment: 29-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Single-use
PES
20 mL
12/Pk.
14 mL
Ultrafiltration
3 kDa
Non-sterile
Centrifugal Concentrator
116 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number251100657 Certificate TypeCertificate of Quality Date11/04/2025 Catalog NumberVS2091
Lot Number241001077 Certificate TypeCertificate of Quality Date01/02/2025 Catalog NumberVS2091
Lot Number240900197 Certificate TypeCertificate of Quality Date17/12/2024 Catalog NumberVS2091
Lot Number235001047 Certificate TypeCertificate of Quality Date16/05/2024 Catalog NumberVS2091
Lot Number230300467 Certificate TypeCertificate of Compliance Date13/03/2024 Catalog NumberVS2091
Special Offers

Special Offers

  • Multiple Manufacturers
    Advance your proteomics research with essential tools
    Promo Code: 25977

    Expires2025-05-31

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sartorius Vivaspin™ 20 Centrifugal Concentrator > Vivaspin 20: PES; 3,000 MWCO; 12/Pk.

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.