Learn More
Socorex™ Acura™ 12-channel Micropipette Volumetric Module
Increase the versatility of your Acura™ electronic micropipette control unit and save costs by accessorizing it with one of 27 available volumetric modules. Socorex™ Acura™ 12-channel Micropipette Volumetric Module is an excellent and easily attachable/detachable choice for your multi-channel micropipetting needs.
Brand:  Socorex™ 800.12.050
 View more versions of this product
Product Code. 10589482
  • £539.00 / Each
Estimated Shipment: 07-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • One of 27 available volumetric assemblies.
    • Increase versatility and save costs.
  • Disassembles and reassembles without tools.
  • Calibration specificities of extra modules - as set by factory QC or by pipette owner - are retained in instrument memory.
Specifications
Specifications
12 Channel Pipette
Acura
50 μL
Acura™ Electro 926 Micropipette
12
Micropipette Volumetric Module
12 Channel Pipette
1 Each
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Socorex™ Acura™ 12-channel Micropipette Volumetric Module > Volume: 50uL