missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford WaterBath Clear Algae Inhibitor
WaterBath Clear contains an active organic ingredient that inhibits Algae growth in water baths and incubators.
Brand: Melford 200120
This item is not returnable.
View return policy
Description
WaterBath Clear contains an active organic ingredient that inhibits Algae growth in water baths and incubators.
WaterBath Clear contains an active organic ingredient that inhibits Algae growth in water baths and incubators. Simply add 0.5ml per liter of water to effectively treat water. Economical to use. Eight ounces treats approximately 500 liters of water. Supplied in a convenient flip cap dispenser bottle.Specifications
Algaecide | |
Clear Liquid | |
8 oz. | |
8 oz. |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 12
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Melford WaterBath Clear Algae Inhibitor > 8 oz.
Spot an opportunity for improvement?Share a Content Correction