missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lenz Laborglasintrumente Weighing Bottles with NS Cap
Click to view available options
Capacity (Metric):
50 mL
Description
- With interchangeable NS cap
- With inscription label
- Available in tall
- Medium and low form
- Made of DURAN™ tubing
Specifications
Specifications
Capacity (Metric) | 50 mL |
Material | Glass |
Height (Metric) | 70 mm |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Lenz Laborglasintrumente Weighing Bottles with NS Cap >
Spot an opportunity for improvement?Share a Content Correction