Learn More
Cytiva Whatman™ 3MM Chr Chromatography Paper
Manufactured entirely from high-quality cotton linters with no additives and tested specifically for chromatographic techniques. Whatman™ Grade 3MM Chr Chromatography Paper ensures the wicking capability and uniformity of capillary action that are important in chemical separations.
£30.10 - £725.00
Specifications
Description 0.34 mm Whatman medium thickness 3 MM Chr paper
Flow Rate 130 mm/30 min.
Film Thickness 0.34 mm
Finish Smooth
For Use With (Application) For Electrophoresis, General Chromatography, and Blotting AppliCatIons
View More Specs
Products 22
Product Code Brand Length (Metric) Width (Metric) Format Price Quantity & Availability  
Product Code Brand Length (Metric) Width (Metric) Format Price Quantity & Availability  
11330744
View Documents Promotion Details
Cytiva
3030-917
57 cm 46 cm Sheet
£481.00
Pack of 100
In Stock
to see stock.
Edge
Add to basket
11895375
View Documents Promotion Details
Cytiva
3030-614
100 m 2 cm Roll
£68.00
Each
In Stock
to see stock.
Add to basket
11348924
View Documents Promotion Details
Cytiva
3030-153
17.5 cm 15 cm Sheet
£79.85
Pack of 100
In Stock
to see stock.
Add to basket
11975194
View Documents
Cytiva
3030-221
34 cm 18 cm Sheet
£125.00
Pack of 100
In Stock
to see stock.
Add to basket
11919197
View Documents Promotion Details
Cytiva
3030-392
45 cm 35 cm Sheet
£267.00
Pack of 100
In Stock
to see stock.
Add to basket
11358934
View Documents Promotion Details
Cytiva
3030-662
100 m 7.5 cm Roll
£183.00
Each
In Stock
to see stock.
Add to basket
10441412
View Documents Promotion Details
Cytiva
3030-672
100 m 10 cm Roll
£300.00
Each
In Stock
to see stock.
Edge
Add to basket
11338934
View Documents
Cytiva
3030-675
100 m 12.5 cm Roll
£231.00
Each
Estimated Shipment: 18-09-2025
to see stock.
Add to basket
11380734
View Documents Promotion Details
Cytiva
3030-681
100 m 15 cm Roll
£365.00
Each
In Stock
to see stock.
Add to basket
11390734
View Documents
Cytiva
3030-690
100 m 19 cm Roll
£452.00
Each
In Stock
to see stock.
Add to basket
11300744
View Documents
Cytiva
3030-700
100 m 23 cm Roll
£389.00
Each
In Stock
to see stock.
Add to basket
11310744
View Documents
Cytiva
3030-704
100 m 27 cm Roll
£475.00
Each
In Stock
to see stock.
Add to basket
11320744
View Documents Promotion Details
Cytiva
3030-861
20 cm 20 cm Sheet
£129.00
Pack of 100
In Stock
to see stock.
Add to basket
10211031
View Documents Promotion Details
Cytiva
3030-931
68 cm 58 cm Sheet
£725.00
Pack of 100
In Stock
to see stock.
Add to basket
11378924
View Documents Promotion Details
Cytiva
3030-866
25 cm 20 cm Sheet
£148.00
Pack of 100
In Stock
to see stock.
Add to basket
11929197
View Documents
Cytiva
3030-6132
14 cm 12 cm Sheet
£33.20
Pack of 100
In Stock
to see stock.
Add to basket
11939197
View Documents
Cytiva
3030-6185
14 cm 11 cm Sheet
£43.50
Pack of 100
In Stock
to see stock.
Add to basket
11328934
View Documents
Cytiva
3030-6187
15.2 cm 20.3 cm Sheet N/A N/A N/A
11949197
View Documents Promotion Details
Cytiva
3030-6188
20 cm 15 cm Sheet
£74.95
Pack of 100
In Stock
to see stock.
Add to basket
10154153
View Documents Promotion Details
Cytiva
3030-6189
10.16 cm 13.34 cm Sheet
£30.10
Pack of 100
In Stock
to see stock.
Add to basket
11955477
View Documents
Cytiva
41 cm 26 cm Sheet
£209.00
Pack of 100
In Stock
to see stock.
Add to basket
11948915
View Documents
Cytiva
3030-347
43 cm 35 cm Sheet
£288.00
Pack of 100
In Stock
to see stock.
Aggiungi al carrello
Descrizione
Descrizione
Manufactured entirely from high-quality cotton linters with no additives and tested specifically for chromatographic techniques. Whatman™ Grade 3MM Chr Chromatography Paper ensures the wicking capability and uniformity of capillary action that are important in chemical separations.
  • Smooth surface
  • No “paper making” additives—the final organic purity of these chromatography grade papers represents the highest level that can be achieved without oxidation and degradation of the cellulose fibers
  • Flowrate: 130mm/30 min. (water)
  • Medium thickness: 0.34mm
  • Widely used for electrophoresis, general chromatography, and blotting applications
Specifica
Specifica
0.34 mm Whatman medium thickness 3 MM Chr paper
0.34 mm
For Electrophoresis, General Chromatography, and Blotting AppliCatIons
Cellulose
130 mm/30 min.
Smooth
3 MM Chr
Videos
SDS
Documenti
Documenti
Certificati
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Cytiva Whatman™ 3MM Chr Chromatography Paper
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato