Learn More
DWK Life Sciences WHEATON™ CompletePAK™ Clear Kits
The WHEATON™ CompletePAK™ product line is a range of crimp top vials, rubber stoppers and seals that are sterilized ready-to-use and are off- the-shelf, and can be customized for any application reducing your supply chain due to our one source solution capability.
Brand:  DWK Life Sciences W991007-CP
1301.06 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 17942631
  • £1405.00 / 1 set
Estimated Shipment: 04-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Sterilized, off-the-shelf, ready-to-use (RtU) products
  • Variety of Kitted and stand-alone sterile components – a solution for any application
  • Kitted solutions feature equal amounts of RtU components for reduced scrap rates
  • Complete manufacturing & certificate of analysis traceability
  • USP 788/EP 2.9.19 Particulate Matter in Injections, USP 85/EP 2.6.1, The Bacterial Endotoxins Test and USP 71/ EP 2.6.14 The Sterility Test, USP 660/EP 3.2.1 Glass Containers for Pharmaceutical Use, and USP 381/EP 3.2.9 Elastomeric Closures used in Parenteral Preparations
  • CompletePAK™ ModPAK Bulk configurations
  • Ready-to-Use, Just-in-Time, minimum 3 month shelf life from time of shipment
  • Container Closure Integrity (CCI) certification available upon request
Certifications
USP <788>, <85> and <71>; Container Closure Integrity (CCI) certified
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Blue
Glass
5 mL
120/Cs.
OmniFlex 3G Serum Stopper
20 mm
Clear
Sterile
Vial, Stopper, and Seal
Wheaton CompletePAK
USP 788, 85, and 71, Container Closure Integrity (CCI) certified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
DWK Life Sciences WHEATON™ CompletePAK™ Clear Kits > 5 mL Clear Sterile Vial (200), 20 mm OmniFlex 3G Sterile Serum Stopper (220), Sterile Blue Seal (220), 120/Cs.