Learn More
DWK Life Sciences Wheaton™ Crimpers
With ergonomically designed handles for sealing standard laboratory vials
Brand: DWK Life Sciences W225301
This item is not returnable.
View return policy
Description
- Wheaton E-Z Crimpers feature cushioned ergonomic handles that aid in reducing hand fatigue and add a high degree of comfort for the user. Each crimper size is labeled for quick identification. Other features include polished crimping jaws for consistent sealing, a short handle and redesigned spring assembly that further aids in reducing hand fatigue. For semi-automatic crimping, use the E-Z Crimper with the Wheaton Crimpmaster™ Crimping Station. An adjustable stop allows you to determine the tightness of the seal while maintaining consistent crimping. Tapered jaws provide for easier vial access when using racks. The 11 mm E- Z Crimper is supplied with a plunger insert to give the option of crimping the top of a vial creating an extra tight seal. We recommend this only for standard opening vials.
- Crimpers can be autoclaved, although autoclaving may change the appearance or reduce product life. They should not be autoclaved at greater than 121°C
Services
Two years
Specifications
Case of 1 | |
11 mm Seals |
WHE026 Catalogue 2012 FINAL LOWRES
File size: 14.8 MB
WHE026 Catalogue 2012 FINAL LOWRES
File size: 14.8 MB
Product Content Correction
Product Title
DWK Life Sciences Wheaton™ Crimpers > Seal Crimper; Attaches 11mm Seals
Spot an opportunity for improvement?Share a Content Correction