missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Stoppers for Serum Bottles and Vials: 2-Leg Stoppers
Provide airtight covers
Brand: DWK Life Sciences 224100-192
This item is not returnable.
View return policy
Description
Provide airtight covers
- Flange-type, straight plug stoppers are secured with aluminum crimp seals (06-406-11B)
- Three-prong plug stoppers are for freeze-dry applications (06-447E, 06-451-56)

Specifications
13 mm | |
Lyophilization | |
50 Durometer | |
Gray | |
Chlorobutyl Isoprene Blend |
20 mm | |
Fits 13mm I.D., 20mm O.D. mouth of serum bottles and vials | |
Autoclavable | |
13 mm | |
2-Leg Lyophilization Stopper; Chlorobutyl Isoprene blend |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
WHE026 Catalogue 2012 FINAL LOWRES
File size: 14.8 MB
WHE026 Catalogue 2012 FINAL LOWRES
File size: 14.8 MB
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DWK Life Sciences Wheaton™ Stoppers for Serum Bottles and Vials: 2-Leg Stoppers > 2-leg Lyophilization Plug; Gray Chlorobutyl-Isoprene blend/50; I.D. x O.D.: 13 x 20mm
Spot an opportunity for improvement?Share a Content Correction