missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Woodsafe™ Woodsafe Sharpsbin Holders
Accessories to Woodsafe sharpsbins and clinical waste bins. Holders and trolleys.
Brand: Woodsafe™ 5500078
141.55 GBP valid until 2025-05-30
Use promo code "24004" to get your promotional price.
This item is not returnable.
View return policy
Description
- Woodsafe™ offer a sustainable clinical waste management solution produced with residue from the forest industry
- Lowers the carbon footprint by more than 60%, proved in full LCA
- Steady holders and trolleys
Specifications
Woodsafe Sharpsbin Holders | |
25 L and 50 L Woodsafe containers | |
Trolley |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 12
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Woodsafe™ Woodsafe Sharpsbin Holders > 25 L and 50 L Woodsafe containers, Trolley
Lower climate impact and ACT label
Spot an opportunity for improvement?Share a Content Correction