missing translation for 'onlineSavingsMsg'
Learn More
1,1'-Diethyl-2,2'-cyanine iodide, 99%, Thermo Scientific Chemicals Product Code.: 10035940

1,1'-Diethyl-2,2'-cyanine iodide, 99%, Thermo Scientific Chemicals

Product Code. 10035940
500 mg, Glass bottle
Click to view available options
Quantity:
500 mg
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10035940

Brand: Thermo Scientific Acros 407255000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 977-96-8
CAS Min % 98.5
CAS Max % 100.0
Melting Point 273°C
Color Red to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C23H23IN2
MDL Number MFCD00011971
Quantity 500 mg
Beilstein 23, II, 267
Synonym decynium 22
InChI Key GMYRVMSXMHEDTL-UHFFFAOYSA-M
SMILES CCN1C(=CC2=[N+](C3=CC=CC=C3C=C2)CC)C=CC4=CC=CC=C41.[I-]
IUPAC Name (2Z)-1-ethyl-2-[(1-ethylquinolin-1-ium-2-yl)methylidene]quinoline;iodide
Molecular Weight (g/mol) 454.34
PubChem CID 71299759
ChEBI CHEBI:37993
Formula Weight 454.34
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 1,1′-Diethyl-2,2′-cyanine iodide, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0442417 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number407255000
Lot NumberA0438271 Certificate TypeCertificate of Analysis Date27/02/2023 Catalog Number407255000
Lot NumberA0371942 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number407255000
Lot NumberA0442417 Certificate TypeCertificate of Analysis Date08/06/2022 Catalog Number407255000
Lot NumberA0431328 Certificate TypeCertificate of Analysis Date17/06/2021 Catalog Number407255000
compliance-icons
Product Identifier
  • 1,1``-Diethyl-2,2``-cyanine iodide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H330-Fatal if inhaled.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,1'-Diethyl-2,2'-cyanine iodide, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.