missing translation for 'onlineSavingsMsg'
Learn More
4'-Iodoacetophenone, 98%, Thermo Scientific Chemicals Product Code.: 10415755

4'-Iodoacetophenone, 98%, Thermo Scientific Chemicals

Product Code. 10415755
5 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10415755

Brand: Thermo Scientific Acros 278890050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13329-40-3
Molecular Formula C8H7IO
Molecular Weight (g/mol) 246.05
MDL Number MFCD00045320
InChI Key JZJWCDQGIPQBAO-UHFFFAOYSA-N
Synonym 4'-iodoacetophenone, 1-4-iodophenyl ethanone, 4-iodoacetophenone, 1-4-iodophenyl ethan-1-one, p-iodoacetophenone, ethanone, 1-4-iodophenyl, 1-4-iodo-phenyl-ethanone, 1-acetyl-4-iodobenzene, 4-iodo acetophenone, 4;-iodoacetophenoneShow MoreShow Less
PubChem CID 72869
IUPAC Name 1-(4-iodophenyl)ethanone
SMILES CC(=O)C1=CC=C(C=C1)I

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 82.0°C to 89.0°C
Color Brown
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula IC6H4C(O)CH3
Quantity 5 g
Formula Weight 246.05
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4'-Iodoacetophenone, 98%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0427522 Certificate TypeCertificate of Analysis Date11/09/2023 Catalog Number278890250,278890050
Lot NumberA0440881 Certificate TypeCertificate of Analysis Date18/04/2022 Catalog Number278890250,278890050
Lot NumberA0404273 Certificate TypeCertificate of Analysis Date19/07/2021 Catalog Number278890250,278890050
Lot NumberA0390699 Certificate TypeCertificate of Analysis Date13/05/2020 Catalog Number278890250,278890050
compliance-icons
Product Identifier
  • 4'-Iodoacetophenone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4'-Iodoacetophenone, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.