missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Accu20 Ultrapure Water System

Fisherbrand™ Accu20 Ultrapure Water System used to produce Type I ultrapure water and RO water on demand from tap water directly.

Brand:  Fisherbrand™ RD0A040UEK

4724.40 GBP valid until 2025-04-30
Use promo code "24119" to get your promotional price.


 View more versions of this product

Product Code. 15499689

  • £8475.00 / Each
Estimated Shipment: 21-10-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Compact system provides easy way to produce ultrapure water or RO water on demand from tap water directly with no additional pre-treatment filters
  • Equipped with safeguard AccuDuo P pack cartridge and dual-column AccuDuo U Pack ultrapurification cartridge to ensure optimum quality of final product water
  • No external storage tank required, which eliminate risk of water degration by contaminants from container or tubing
  • Fully automatic controlled and maintained system contains automatic cleaning cycles and selfmaintenance functions for RO membrane, which increases lifetime and efficiency of RO membranes as well as cartridges
  • Automatic hourly system recirculation in loop prevents bacterial growth and maintain water purity at all times
  • System status and key quality parameters are clearly displayed on large LCD screen for easy system performance monitoring

Applications:

  • HPLC mobile phase preparation
  • Preparation of reagent blank solutions
  • As sample diluent for GC, HPLC, AA, ICP-MS and other analytical techniques
  • Preparation of buffers and culture media for mammalian cell culture
  • Preparation of molecular biology reagents, etc.

Certifications

Meets or exceeds ASTM, CLSI, CAP, and ISO Type I water standards

Specifications

Specifications

RO water at 25°C: Typically <20μS/cm
RO water (at 25°C ): >0.5L/min.; Ultrapure water ( Type I ) (at 25°C ): >0.5L/min.
<10 ppb (<5 ppb with a dual wavelength UV lamp )
15 to 90 psi (1 to 6 bar )
<1/mL ( with a 0.2μm final filter or terminal ultrafiltration cartridge )
76.2 in.
18.8 in.
20 in.
<0.1 CFU/mL (with a 0.2μm final filter or terminal ultrafiltration cartridge)
18.2 Mohm.cm
ACCU20 UV Ultrapure Water System
Accu20
5°C to 45°C
30 cm
48 cm
51 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Accu20 Ultrapure Water System > ACCU20 UV Ultrapure water system

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.