Learn More
Azlon™ Silicone Rubber Hand/Finger Protector Product Code.: 12008841
Azlon™ Silicone Rubber Hand/Finger Protector
Product Code. 12008841
Small
Click to view available options
Size:
Finger
Medium
Small
Product Code. 12008841
Brand: Azlon PGR404
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Ideal for safe handling of hot beakers, flasks
  • Capable of withstanding temperatures up to 250°C
  • For handling cold objects down to -57°C
  • Contact surfaces are multi-studded for positive grip
TRUSTED_SUSTAINABILITY
Specifications
Material Silicone
Product Type Hand/Finger Protector
Size Small
For Use With (Application) For Safe Handling Of Hot Beakers, Flasks etc
Product Content Correction
Product Title
Azlon™ Silicone Rubber Hand/Finger Protector >