missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sperian™ Honeywell™ ARATHERMA FIT™ Thermal Protection Gloves
Click to view available options
Size:
9
Description
- Kevlar™-lined glove with cotton is pleasant to the touch
- Lining enhances the glove's mechanical performance while absorbing perspiration
- Ambidextrous and thus more cost-effective
- Glove's relative thickness (gauge 7) enables it to absorb shock, making it suitable for handling heavy objects
- Kevlar fiber offers good resistance to cuts and protects against thermal hazards
- Length of the knitted wrist offers forearm protection
- Provide protection for Level 2 heat exposure, which is effective up to 250°C

Spécification
Spécification
Certifications/Compliance | EC 0075/797/162/06/08/0191 |
Classification | CE CATEGORY II |
Coating Material | Dupont™ Kevlar™ aramid fibers |
Cut Resistance Rating | 5 |
Hand | Ambidextrous |
Product Type | Gloves |
Size | 9 |
Temperature (Metric) Max | Effective up to 250°C |
Liner Material | Cotton |
Quantity | 1 Each |
Afficher plus |
Suggestions de produits
Customers who viewed this item also viewed
Viewing 1-5 of 15
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Sperian™ Honeywell™ ARATHERMA FIT™ Thermal Protection Gloves >
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu