missing translation for 'onlineSavingsMsg'
Learn More
Benzyl isocyanate, 99+%, Thermo Scientific Chemicals Product Code.: 10286550

Benzyl isocyanate, 99+%, Thermo Scientific Chemicals

Product Code. 10286550
5 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10286550

Brand: Thermo Scientific Acros 210760050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3173-56-6
Molecular Formula C8H7NO
Molecular Weight (g/mol) 133.15
MDL Number MFCD00009701
InChI Key YDNLNVZZTACNJX-UHFFFAOYSA-N
Synonym benzyl isocyanate, isocyanatomethyl benzene, benzylisocyanate, isocyanic acid benzyl ester, benzene, isocyanatomethyl, 1-isocyanatomethyl benzene, phenylmethyl isocyanate, unii-8i2li357gq, phenylmethanisocyanateShow MoreShow Less
PubChem CID 76639
IUPAC Name isocyanatomethylbenzene
SMILES O=C=NCC1=CC=CC=C1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0700g/mL
Boiling Point 101°C to 104°C (33.0 mmHg)
Flash Point 45°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula C6H5CH2NCO
Refractive Index 1.5241 to 1.5261
Quantity 5 g
Beilstein 12, 1059
Fieser 13,30; 01,1005
Specific Gravity 1.07
Formula Weight 133.15
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Benzyl isocyanate, 99+%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0468280 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number210760250,210760050
Lot NumberA0456960 Certificate TypeCertificate of Analysis Date14/02/2024 Catalog Number210760250,210760050
Lot NumberA0397514 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number210760250,210760050
Lot NumberA0407760 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number210760250,210760050
Lot NumberA0445747 Certificate TypeCertificate of Analysis Date13/09/2022 Catalog Number210760250,210760050
compliance-icons compliance-icons
Product Identifier
  • Benzyl isocyanate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Benzyl isocyanate, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.