Learn More
Jaytec Glass™ Borosilicate Glass Butyrometer Tube
The Borosilicate Glass Butyrometer Tube is used to determine the fat content of dairy samples using the Gerber method.
Brand: Jaytec Glass™ MP893
Description
- Borosilicate glass
- Capacity and tolerances in accordance with BS ISO 488:2008 (formerly BS 696)
- Graduated round stem
Specifications
Borosilicate Glass | |
Milk Butyrometer | |
0.1% |
10.94 mL | |
Determination of fat content using Gerber method | |
0 to 8% |