missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Brilliance™ UTI Clarity

Obtain fast and accurate results with a chromogenic medium designed for screening clinical samples for the presence of common urinary tract pathogens.

Brand:  Thermo Scientific™ PO1110A

30.24 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 12967198

  • £58.10 / Pack of 10

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Enumerate, identify and isolate common organisms associated with urinary tract infections within 18 to 24 hours using Thermo Scientific™ Brilliance™ UTI Clarity with no further confirmation for E.coli. Organisms are differentiated through the utilization of one or more chromogens and/or tryptophan. Interpretation is aided by the inclusion of an transparent background, providing a distinct contrast against the seven possible colors of colony growth.

The Brilliance UTI Clarity formulation contains two chromogenic substrates which are cleaved by the ß-galactosidase and ß-glucosidase enzymes produced by E. coli, Enterococcus species and coliforms. These specific enzyme reactions cleave the chromogens giving a range of diagnostic colors.

The ß-galactosidase activity of E. coli and S. saprophyticus results in pink/red colonies and the ß-glucosidase activity of enterococci produces blue/turquoise colonies; the activity of both enzymes on coliforms gives dark blue/purple colonies. Tryptophan deaminase activity produces a brown halo around colonies of Proteus, Morganella and Providencia species. Most other organisms exhibit their natural pigmentation.

The specificity of Brilliance UTI clarity minimises confirmatory testing, with same day results for E.coli confirmation being achieved using a rapid indole test (DMAC) (MB1448A).

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Straw, clear
90mm Monoplate
Brilliance UTI Clarity
19.5g ± 1.0g
10/Pk.
2°C to 10°C
United Kingdom
Agar
Flow Wrapped - Single Layer
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number4504029 Certificate TypeCertificate of Analysis Date16/04/2025 Catalog NumberPO1110A
Lot Number4503256 Certificate TypeCertificate of Analysis Date14/04/2025 Catalog NumberPO1110A
Lot Number4503436 Certificate TypeCertificate of Analysis Date08/04/2025 Catalog NumberPO1110A
Lot Number4501377 Certificate TypeCertificate of Analysis Date07/04/2025 Catalog NumberPO1110A
Lot Number4500765 Certificate TypeCertificate of Analysis Date07/04/2025 Catalog NumberPO1110A
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Brilliance™ UTI Clarity >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.