Learn More
Fisherbrand™ Bunsen Burner Neoprene Tubing
Suitable for use with coal and natural (methane) gas burners.
Brand:  Fisherbrand™ FB50829
 View more versions of this product
Product Code. 11875522
  • £32.35 / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
With reinforced ends for fitting to Bunsen burner and gas tap
Specifications
Specifications
Neoprene
900 mm
1.5 mm
Bunsen Burner
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Bunsen Burner Neoprene Tubing > Length: 900mm