missing translation for 'onlineSavingsMsg'
Learn More

Cell Culture Grade Water, Corning™
BIOPHARMA

CORNING™ Cell Culture Grade Water is tested to USP/EP sterile water for injection specifications.

Brand:  Corning 25-065-LB

426.49 GBP valid until 2025-05-31
Use promo code "24127" to get your promotional price.


Product Code. 15676675

  • £448.94 / 20 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • High-quality water suitable for use as a solvent in the preparation of cell culture media and other reagents for sensitive applications.
  • Our bioprocess container selections are suitable for large-scale applications such as cleaning and upstream and downstream reagent preparation.
  • Cell culture grade water goes through a wide array of testing which includes testing to the chemical and physical properties found in the USP and EP monographs for sterile Water for Injection (WFI).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Bag
≤3mg (0.0003%)
Sterile
20 L
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cell Culture Grade Water, Corning™ > 20L

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.