missing translation for 'onlineSavingsMsg'
Learn More

Heathrow Scientific™ True North™ Mini-Cooler

The True North™ Mini Cooler protects temperature-sensitive samples at the bench or in the freezer. The Mini Cooler is ideal for keeping PCR™ reagents, enzymes, and other biochemical reagents cool while working at the bench.

Brand:  Heathrow Scientific™ HS120073

116.08 GBP valid until 2025-04-30
Use promo code "24119" to get your promotional price.


 View more versions of this product

Product Code. 16320686

  • £214.00 / Each
Estimated Shipment: 21-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Polycarbonate construction
  • Cooler has rubber non-slip feet
  • Gridded and keyed one-position lid for easy sample identification
  • Available with lid filled with insulating gel
Specifications

Specifications

Blue
3.9 in.
5.7 in.
4.1 in.
Tubes
1 Ea.
Polycarbonate
0.5 to 2 mL
10 cm
14.5 cm
10.5 cm
Clear Lid
Tubes
3 x 4
Cooler
-20°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Heathrow Scientific™ True North™ Mini-Cooler > Color: Blue; Wells: 12; Lid: Clear

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.